Would you like to log in again?
Electrothermal CMUA Series Stirring, Heating Mantles, Cole-Parmer
Supplier: Cole Parmer
Electrothermal
36013-08 36013-13 36013-11
75994-962EA 1898.04 USD InStock
75994-962 75994-960 75994-964
Electrothermal CMUA Series Stirring, Heating Mantles, Cole-Parmer
Heaters Heating Mantles
Magnetic stirrer and heating mantle in one convenient unit.
  • Flexible coiled heating element wraps the heater around round-bottom flask to optimize heat transfer
  • 'Cool to the touch' exterior provides safe handling
These stirring, heating mantles combine the functionality of a heating mantle with a magnetic stirrer providing even heating and mixing. Element temperatures can reach up to 825 °F (450 °C) yet the exterior remains cool to the touch. For maximum safety, the flask does not come in direct contact with the heating element.
Built-in energy regulator and safety earth (ground) screen offer added safety protection. Reach stirring speeds up to 2000 rpm. Simply use the auto-recapture switch to reactivate stirring if stirring is interrupted and stirring motion is lost. Two colored lights by each control dial indicate 'power on' and 'heat on'. Powder-coated aluminum housing makes it easy to clean.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty