Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2'-O-Methyladenosine 98%

Supplier: Ambeed
Warning
Synonyms: (2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)-4-methoxytetrahydrofuran-3-ol

A362743-250MG A362743-25G A362743-100MG A362743-1G A362743-5G
103837-344EA 98 USD InStock
103837-344 103826-328 77003-390 77003-392 77572-074
2'-O-Methyladenosine 98%
2'-O-Methyladenosine
Caution: for Research Use Only


Formula: C₁₁H₁₅N₅O₄
MW: 281.27 g/mol
Boiling Pt: 623.8 °C (760 mm Hg)
Melting Pt: 200…202 °C
Flash Pt: 331 °C
Storage Temperature: Freezer
MDL Number: MFCD00056002
CAS Number: 2140-79-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty