Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-isoleucine 97%

Supplier: Ambeed
Warning
Synonyms: Ac-Ile-OH(2S,3S)-2-Acetamido-3-methylpentanoic acid

A533721-5G A533721-10G
77116-744EA 12.36 USD InStock
77116-744 103839-700
N-Acetyl-L-isoleucine 97%
N-Acetyl-L-isoleucine
Caution: for Research Use Only


Formula: C₈H₁₅NO₃
MW: 173.212 g/mol
Boiling Pt: 369.7 °C (760 mm Hg)
Flash Pt: 177.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00066072
CAS Number: 3077-46-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty