Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-Ethyl-N-boc-pyroglutamate 97%

Supplier: Ambeed
Warning
Synonyms: Boc-Pyr-OEt(S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid ethyl ester(S)-Ethyl-N-Boc-pyroglutamateN-Boc-L-pyroglutamic acid ethyl esterEthyl N-Boc-L-pyroglutamate(S)-1-Boc-5-oxopyrrolidine-2-carboxylic acid ethyl esterEthyl (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate1-Boc-L-Pyroglutamic acid ethyl esterEthyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylateEthyl N-(tert-Butoxycarbonyl)-L-pyroglutamateN-(tert-Butoxycarbonyl)-L-pyroglutamic acid ethyl ester

A213959-100G A213959-5G A213959-500G A213959-10G A213959-5G A213959-25G A213959-1000G
76999-698EA 26.78 USD InStock
76999-698 77303-312 76999-696 76999-702 76999-700 103849-064 77087-564
(S)-Ethyl-N-boc-pyroglutamate 97%
(S)-Ethyl-N-boc-pyroglutamate
Caution: for Research Use Only


Formula: C₁₂H₁₉NO₅
MW: 257.3 g/mol
Boiling Pt: 375 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07778090
CAS Number: 144978-12-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty