Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Hydroxymethyl)benzyl alcohol 99% (by HPLC)

Supplier: Ambeed
Warning
Synonyms: 1,4-Bis(hydroxymethyl)-benzenep-Xylene-α,α'-diolp-Phenylenedimethanol1,4-Benzenedimethanol

A123906-1000G A123906-5G A123906-100G A123906-25G A123906-500G A123906-1000G A123906-10G
103808-286EA 123 USD InStock
103808-286 77189-788 77189-796 77189-792 77189-794 77312-584 77189-790
4-(Hydroxymethyl)benzyl alcohol 99% (by HPLC)
4-(Hydroxymethyl)benzyl alcohol
Caution: for Research Use Only


Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 328.9 °C (760 mm Hg)
Melting Pt: 114…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00004665
CAS Number: 589-29-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty