Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-3-chlorobenzene 98%

Supplier: Ambeed
Warning
Synonyms: 3-Bromochlorobenzene

A145019-25G A145019-5G
103841-564EA 5 USD InStock
103841-564 77316-836
1-Bromo-3-chlorobenzene 98%
1-Bromo-3-chlorobenzene
Caution: for Research Use Only


Formula: C₆H₄BrCl
MW: 191.45 g/mol
Boiling Pt: 196 °C (760 mm Hg)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00000568
CAS Number: 108-37-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty