Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Methylcyclopentanol 97%

Supplier: Ambeed
Danger

A144549-1G A144549-25G A144549-10G A144549-1G A144549-5G A144549-5G
77316-750EA 12 USD InStock
77316-750 103803-420 103841-496 77235-140 77316-748 77299-398
1-Methylcyclopentanol 97%
1-Methylcyclopentanol
Caution: for Research Use Only


Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 137.1 °C (760 mm Hg)
Melting Pt: 36…37 °C
Flash Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD00001364
CAS Number: 1462-03-9
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty