Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Benzyloxy)benzaldehyde 97%

Supplier: Ambeed
Warning

A125047-100G A125047-10G A125047-5G A125047-25G A125047-1000G A125047-500G
77258-230EA 13 USD InStock
77258-230 77258-228 103808-306 77258-226 77258-234 77258-232
2-(Benzyloxy)benzaldehyde 97%
2-(Benzyloxy)benzaldehyde
Caution: for Research Use Only


Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 326 °C (760 mm Hg)
Melting Pt: 46…47 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00016583
CAS Number: 5896-17-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty