Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dibromothiophene 98%

Supplier: Ambeed
Danger

A665351-5G A665351-25G A665351-10G A665351-5G
103840-466EA 8.08 USD InStock
103840-466 77293-102 76844-666 76844-664
2,3-Dibromothiophene 98%
2,3-Dibromothiophene
Caution: for Research Use Only


Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 221.9 °C (760 mm Hg)
Flash Pt: 51.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005418
CAS Number: 3140-93-0
UN: 1992
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty