Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dichloroquinoxaline 98%

Supplier: Ambeed
Warning

A204235-100G A204235-500G A204235-5G A204235-10G A204235-25G A204235-5G
77248-900EA 251 USD InStock
77248-900 77248-898 77086-360 103848-198 77086-358 77248-896
2,3-Dichloroquinoxaline 98%
2,3-Dichloroquinoxaline
Caution: for Research Use Only


Formula: C₈H₄Cl₂N₂
Boiling Pt: 269.7 °C (760 mm Hg)
Melting Pt: 152…154 °C
Flash Pt: 142.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00006720
CAS Number: 2213-63-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty