Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Difluoro-4-hydrazinopyridine 97%

Supplier: Ambeed
Warning
Synonyms: 2,6-Difluoro-4-hydrazinylpyridine

A101284-5G A101284-1G A101284-1G
103795-892EA 465.23 USD InStock
103795-892 103821-578 77297-044
2,6-Difluoro-4-hydrazinopyridine 97%
2,6-Difluoro-4-hydrazinopyridine
Caution: for Research Use Only


Formula: C₅H₅F₂N₃
MW: 145.11 g/mol
Boiling Pt: 255.6±40.0 °C (760 mm Hg)
Melting Pt: 200…202 °C
Storage Temperature: Freezer
MDL Number: MFCD08436095
CAS Number: 837364-94-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty