Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-N-methylacetamide 97%

Supplier: Ambeed
Danger

A153691-5G A153691-500G A153691-5G A153691-10G A153691-25G
77110-416EA 245.14 USD InStock
77110-416 77300-022 77110-414 103842-728 77300-024
2-Chloro-N-methylacetamide 97%
2-Chloro-N-methylacetamide
Caution: for Research Use Only


Formula: C₃H₆ClNO
MW: 107.54 g/mol
Melting Pt: 38…40 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00018913
CAS Number: 96-30-0
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty