Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-4-nitroaniline 97%

Supplier: Ambeed
Warning

A150827-100G A150827-25G A150827-5G A150827-10G A150827-1G A150827-5G
76973-094EA 11.33 USD InStock
76973-094 76973-096 76973-092 103842-342 77317-874 77317-876
2-Fluoro-4-nitroaniline 97%
2-Fluoro-4-nitroaniline
Caution: for Research Use Only


Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Boiling Pt: 317.2 °C (760 mm Hg)
Melting Pt: 126…130 °C
Flash Pt: 145.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00034560
CAS Number: 369-35-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty