Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitrobenzenesulfonamide 98%

Supplier: Ambeed
Warning

A125850-100G A125850-10G A125850-500G A125850-5G
77108-826EA 63.86 USD InStock
77108-826 77108-828 103831-586 77108-830
2-Nitrobenzenesulfonamide 98%
2-Nitrobenzenesulphonamide
Caution: for Research Use Only


Formula: C₆H₆N₂O₄S
MW: 202.19 g/mol
Boiling Pt: 418.8 °C (760 mm Hg)
Melting Pt: 189…194 °C
Flash Pt: 207.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00009807
CAS Number: 5455-59-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty