Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(2-Methoxyphenyl)propionic acid 98%

Supplier: Ambeed
Warning

A461673-5G A461673-25G A461673-1G A461673-5G A461673-1G
77307-432EA 16 USD InStock
77307-432 77307-430 103839-008 76842-894 103827-932
3-(2-Methoxyphenyl)propionic acid 98%
3-(2-Methoxyphenyl)propionic acid
Caution: for Research Use Only


Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 305 °C (760 mm Hg)
Melting Pt: 85…87 °C
Flash Pt: 119.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00002772
CAS Number: 6342-77-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty