Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Trifluoromethyl)benzylamine 98%

Supplier: Ambeed
Danger

A233117-10G A233117-5G A233117-25G A233117-5G
103850-630EA 12 USD InStock
103850-630 77089-580 77089-578 77249-610
3-(Trifluoromethyl)benzylamine 98%
3-(Trifluoromethyl)benzylamine
Caution: for Research Use Only


Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 180.6 °C (760 mm Hg)
Flash Pt: 71.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008117
CAS Number: 2740-83-2
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty