Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dibromosalicylaldehyde 98%

Supplier: Ambeed
Warning
Synonyms: 3,5-Dibromo-2-hydroxybenzaldehyde

A622529-5G A622529-500G A622529-25G
76844-388EA 19.57 USD InStock
76844-388 103840-222 77117-470
3,5-Dibromosalicylaldehyde 98%
3,5-Dibromosalicylaldehyde
Caution: for Research Use Only


Formula: C₇H₄Br₂O₂
MW: 279.92 g/mol
Melting Pt: 82…83.5 °C
Flash Pt: 111.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00003318
CAS Number: 90-59-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty