Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichlorobenzonitrile 97%

Supplier: Ambeed
Warning

A340190-25G A340190-5G A340190-5G A340190-10G
76839-802EA 9.27 USD InStock
76839-802 76839-800 103836-784 77115-004
3,5-Dichlorobenzonitrile 97%
3,5-Dichlorobenzonitrile
Caution: for Research Use Only


Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
Boiling Pt: 223.279 °C (760 mm Hg)
Melting Pt: 64…67 °C at N/A mmHg
Flash Pt: 85.849 °C
Storage Temperature: Ambient
MDL Number: MFCD00001800
CAS Number: 6575-00-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty