Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,6-Dibromocarbazole 97%

Supplier: Ambeed
Warning
Synonyms: 3,6-Dibromo-9H-carbazole

A206018-100G A206018-1G A206018-10G A206018-500G A206018-5G A206018-5G A206018-25G
76721-568EA 170.98 USD InStock
76721-568 76721-564 103848-352 77086-586 76721-566 77086-588 76721-562
3,6-Dibromocarbazole 97%
3,6-Dibromocarbazole
Caution: for Research Use Only


Formula: C₁₂H₇Br₂N
Boiling Pt: 459 °C (760 mm Hg)
Melting Pt: 204…206 °C
Flash Pt: 231.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004961
CAS Number: 6825-20-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty