Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,6-Dimethyl-1,4-dioxane-2,5-dione 95%

Supplier: Ambeed
Warning
Synonyms: DL-Lactide

A267054-1G A267054-100G A267054-10G A267054-5G A267054-1G A267054-5G A267054-500G A267054-25G
77092-450EA 10.3 USD InStock
77092-450 77304-968 103812-828 76801-914 76801-916 103852-730 76801-918 77092-448
3,6-Dimethyl-1,4-dioxane-2,5-dione 95%
3,6-Dimethyl-1,4-dioxane-2,5-dione
Caution: for Research Use Only


Formula: C₆H₈O₄
MW: 144.13 g/mol
Melting Pt: 116…119 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011685
CAS Number: 95-96-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty