Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzonitrile 98%

Supplier: Ambeed
Warning
Synonyms: m-Bromobenzonitrile

A105614-100G A105614-5G A105614-10G A105614-1000G A105614-500G A105614-25G
77107-560EA 6.18 USD InStock
77107-560 77107-564 77107-562 77107-558 103822-646 103807-936
3-Bromobenzonitrile 98%
3-Bromobenzonitrile
Caution: for Research Use Only


Formula: C₇H₄BrN
MW: 182.02 g/mol
Boiling Pt: 225.8 °C (760 mm Hg)
Melting Pt: 38…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00001796
CAS Number: 6952-59-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty