Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-6-iodopyridazine 97%

Supplier: Ambeed
Warning

A180200-10G A180200-100G A180200-5G A180200-1G A180200-250MG
103807-520EA 11 USD InStock
103807-520 103845-874 77550-162 77274-024 77274-026
3-Chloro-6-iodopyridazine 97%
3-Chloro-6-iodopyridazine
Caution: for Research Use Only


Formula: C₄H₂ClIN₂
MW: 240.43 g/mol
Boiling Pt: 327.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08275187
CAS Number: 135034-10-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty