Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Iodo-p-toluidine 98%

Supplier: Ambeed
Danger

A242319-5G A242319-25G
103851-274EA 16.15 USD InStock
103851-274 77090-446
3-Iodo-p-toluidine 98%
3-Iodo-p-toluidine
Caution: for Research Use Only


Formula: C₇H₈IN
MW: 233.05 g/mol
Boiling Pt: 268.7 °C (760 mm Hg)
Melting Pt: 34…37 °C
Flash Pt: 116.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00047843
CAS Number: 35944-64-0
UN: 2923
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty