Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methoxy-4-methylbenzonitrile 98%

Supplier: Ambeed
Danger

A127069-1G A127069-25G A127069-5G A127069-10G A127069-1G
103831-774EA 19.62 USD InStock
103831-774 77271-828 103800-906 77298-174 77298-172
3-Methoxy-4-methylbenzonitrile 98%
2-Methoxy-5-methylbenzonitrile
Caution: for Research Use Only


Formula: C₉H₉NO
MW: 147.1739 g/mol
Boiling Pt: 247.2 °C (760 mm Hg)
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD03428590
CAS Number: 3556-60-3
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty