Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Nitrophthalonitrile 98%

Supplier: Ambeed
Warning
Synonyms: 3-Nitrobenzene-1,2-dicarbonitrile3-Nitro-1,2-dicyanobenzene3-Nitro-1,2-benzenedicarbonitrile

A197272-1G A197272-10G A197272-1000G A197272-5G A197272-500G A197272-100G A197272-1000G A197272-25G A197272-5G
103847-528EA 6.92 USD InStock
103847-528 76770-028 77085-524 77085-522 77085-520 76770-032 103809-322 76770-030 77550-256
3-Nitrophthalonitrile 98%
3-Nitrophthalonitrile
Caution: for Research Use Only


Formula: C₈H₃N₃O₂
Boiling Pt: 386.1 °C (760 mm Hg)
Melting Pt: 162…165 °C
Flash Pt: 187.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00191558
CAS Number: 51762-67-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty