Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethyl)cyclohexanone 95%

Supplier: Ambeed
Danger

A198873-250MG A198873-5G A198873-1G
103847-680EA 181 USD InStock
103847-680 103817-052 77287-620
4-(Trifluoromethyl)cyclohexanone 95%
4-(Trifluoromethyl)cyclohexanone
Caution: for Research Use Only


Formula: C₇H₉F₃O
MW: 166.14
Boiling Pt: 156.2 °C (760 mm Hg)
Flash Pt: 53.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00102145
UN: 1224
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty