Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorosalicylic acid 98%

Supplier: Ambeed
Warning
Synonyms: 4-Fluoro-2-hydroxybenzoic acid

A168162-10G A168162-5G A168162-25G A168162-100G A168162-500G A168162-1G A168162-1G A168162-5G
103806-258EA 6.92 USD InStock
103806-258 76973-888 77081-568 77081-566 76973-884 77300-920 76973-886 103844-516
4-Fluorosalicylic acid 98%
4-Fluorosalicylic acid
Caution: for Research Use Only


Formula: C₇H₅FO₃
MW: 156.11 g/mol
Boiling Pt: 300.5 °C (760 mm Hg)
Melting Pt: 170 °C
Flash Pt: 135.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00153168
CAS Number: 345-29-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty