Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-7-nitrobenzofurazan 95%

Supplier: Ambeed
Warning
Synonyms: NBD-F4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole

A297151-1G A297151-100MG
77002-436EA 123.6 USD InStock
77002-436 103814-126
4-Fluoro-7-nitrobenzofurazan 95%
4-Fluoro-7-nitrobenzofurazan
Caution: for Research Use Only


Formula: C₆H₂FN₃O₃
MW: 183.1 g/mol
Boiling Pt: 308.6 °C (760 mm Hg)
Melting Pt: 52…54 °C
Flash Pt: 113 °C
Storage Temperature: Freezer
MDL Number: MFCD00010196
CAS Number: 29270-56-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty