Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxythiobenzamide 98%

Supplier: Ambeed
Warning
Synonyms: 4-Hydroxybenzothioamide

A149152-1000G A149152-25G A149152-500G A149152-1000G A149152-100G
77190-722EA 92 USD InStock
77190-722 77190-720 103808-724 77317-614 77317-612
4-Hydroxythiobenzamide 98%
4-Hydroxythiobenzamide
Caution: for Research Use Only


Formula: C₇H₇NOS
MW: 153.2 g/mol
Boiling Pt: 320.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04973332
CAS Number: 25984-63-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty