Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Isopropoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A262574-10G A262574-25G A262574-1G A262574-1G A262574-5G
103812-568EA 10.38 USD InStock
103812-568 77092-084 103852-464 76692-722 77304-848
4-Isopropoxybenzaldehyde 98%
4-Isopropoxybenzaldehyde
Caution: for Research Use Only


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 264.2 °C (760 mm Hg)
Flash Pt: 112.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052357
CAS Number: 18962-05-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty