Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Formylthiophene-2-carbonitrile 98%

Supplier: Ambeed
Warning
Synonyms: 5-Cyanothiophene-2-carboxaldehyde

A101751-10G A101751-25G A101751-100MG A101751-5G A101751-250MG A101751-1G A101751-1G A101751-5G
77308-710EA 36 USD InStock
77308-710 76765-188 76765-186 76765-184 77308-708 103821-800 76765-190 103796-114
5-Formylthiophene-2-carbonitrile 98%
5-Formylthiophene-2-carbonitrile
Caution: for Research Use Only


Formula: C₆H₃NOS
MW: 137.156 g/mol
Boiling Pt: 297.2 °C (760 mm Hg)
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD09037804
CAS Number: 21512-16-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty