Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Nitro-1,10-phenanthroline 98%

Supplier: Ambeed
Warning

A157366-1G A157366-250MG A157366-1G A157366-5G A157366-5G
77319-012EA 16 USD InStock
77319-012 103804-994 103843-196 77319-014 76719-968
5-Nitro-1,10-phenanthroline 98%
5-Nitro-1,10-phenanthroline
Caution: for Research Use Only


Formula: C₁₂H₇N₃O₂
Boiling Pt: 444 °C (760 mm Hg)
Melting Pt: 202…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00004981
CAS Number: 4199-88-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty