Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9,10-Dibromoanthracene 98%

Supplier: Ambeed
Warning

A518857-500G A518857-5G A518857-5G A518857-25G A518857-100G A518857-10G
77307-702EA 6 USD InStock
77307-702 103839-592 76980-000 76979-996 76843-596 76979-998
9,10-Dibromoanthracene 98%
9,10-Dibromoanthracene
Caution: For research use only


Formula: C₁₄H₈Br₂
MW: 336.03 g/mol
Boiling Pt: 427.1 °C (760 mm Hg)
Melting Pt: 222…223 °C
Flash Pt: 248.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00001244
CAS Number: 523-27-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty