Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-D-ABU-OH 97%

Supplier: Ambeed
Warning
Synonyms: (R)-2-(Boc-amino)butyric acid

A250281-10G A250281-5G A250281-1G A250281-1G A250281-25G A250281-500G A250281-5G A250281-100G
76692-468EA 19.57 USD InStock
76692-468 103811-916 77091-102 76692-466 77091-104 103851-754 76692-470 77304-514
Boc-D-ABU-OH 97%
Boc-D-ABU-OH
Caution: for Research Use Only


Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
Boiling Pt: 334.5 °C (760 mm Hg)
Flash Pt: 156.11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270335
CAS Number: 45121-22-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty