Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-D-leucinol 95%

Supplier: Ambeed
Warning
Synonyms: N-Boc-D-leucinol

A201764-1G A201764-5G A201764-1G A201764-10G
77086-050EA 10.3 USD InStock
77086-050 76999-346 103817-326 103847-966
Boc-D-leucinol 95%
Boc-D-leucinol
Caution: for Research Use Only


Formula: C₁₁H₂₃NO₃
MW: 217.309 g/mol
Boiling Pt: 319.3 °C (760 mm Hg)
Flash Pt: 146.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00235931
CAS Number: 106930-51-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty