Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-D-4-Cyanophenylalanine 98%

Supplier: Ambeed
Warning
Synonyms: Boc-D-Phe(4-CN)-OHBoc-4-cyano-D-Phe-OHN-Boc-4-cyano-D-phenylalanine

A100907-250MG A100907-10G A100907-5G A100907-100MG A100907-5G A100907-1G A100907-1G A100907-25G
77296-902EA 84 USD InStock
77296-902 76780-422 77296-900 76780-420 103795-686 76780-418 103821-376 76780-416
Boc-D-4-Cyanophenylalanine 98%
Boc-D-4-Cyanophenylalanine
Caution: for Research Use Only


Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
Boiling Pt: 494 °C (760 mm Hg)
Melting Pt: 152.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672527
CAS Number: 146727-62-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty