Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-N-ethylglycine 97%

Supplier: Ambeed
Warning
Synonyms: N-Ethylglycine, N-BOC protected

A104447-1G A104447-10G A104447-5G A104447-250MG A104447-5G
103796-708EA 7 USD InStock
103796-708 76687-182 77309-290 76687-180 103822-422
N-Boc-N-ethylglycine 97%
N-Boc-N-ethylglycine
Caution: for Research Use Only


Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Boiling Pt: 320.2 °C (760 mm Hg)
Melting Pt: 28 °C
Flash Pt: 136.886 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672508
CAS Number: 149794-10-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty