Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-iodophenyl)propanoic acid 98%

Supplier: Ambeed
Warning
Synonyms: Boc-p-iodo-Phe-OHBoc-Phe(4-I)-OH

A100839-25G A100839-1G A100839-1G A100839-5G A100839-10G A100839-100G A100839-5G
77282-584EA 26 USD InStock
77282-584 76780-404 77296-860 103821-348 76780-408 76780-406 103795-652
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-iodophenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-iodophenyl)propanoic acid
Caution: for Research Use Only


Formula: C₁₄H₁₈INO₄
MW: 391.2 g/mol
Boiling Pt: 475.3 °C (760 mm Hg)
Melting Pt: 150 °C
Flash Pt: 241.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00037119
CAS Number: 62129-44-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty