Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid 98%

Supplier: Ambeed
Warning
Synonyms: Boc-Phe(4-NO2)-OHN-Boc-4-nitro-L-phenylalanineBoc-p-nitro-Phe-OH

A177764-100G A177764-25G A177764-5G A177764-5G A177764-500G A177764-10G
76974-316EA 57.68 USD InStock
76974-316 103845-600 76974-318 76974-320 77082-992 77301-524
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-nitrophenyl)propanoic acid
Caution: for Research Use Only


Formula: C₁₄H₁₈N₂O₆
MW: 310.3 g/mol
Melting Pt: 107 °C
Flash Pt: 261.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00038128
CAS Number: 33305-77-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty