Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Di(2-pyridyl) ketone 97%

Supplier: Ambeed
Warning
Synonyms: Di(pyridin-2-yl)methanone

A107780-5G A107780-1G A107780-500G A107780-25G A107780-10G A107780-5G A107780-100G A107780-1G
76687-314EA 1684.05 USD InStock
76687-314 76687-312 76687-310 103823-014 77283-644 77283-642 76687-308 103797-246
Di(2-pyridyl) ketone 97%
Di(2-pyridyl) ketone
Caution: for Research Use Only


Formula: C₁₁H₈N₂O
MW: 184.20 g/mol
Boiling Pt: 343.5 °C (760 mm Hg)
Melting Pt: 52…55 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006288
CAS Number: 19437-26-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty