Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid 98%

Supplier: Ambeed
Warning
Synonyms: Fmoc-Cit-OHN-Fmoc-L-citrullineNalpha-Fmoc-L-citrulline

A172625-5G A172625-100G A172625-5G A172625-10G A172625-500G A172625-25G
103845-022EA 6 USD InStock
103845-022 77082-228 76720-558 76720-556 76720-560 77301-204
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid
Caution: for Research Use Only


Formula: C₂₁H₂₃N₃O₅
MW: 397.4 g/mol
Melting Pt: 168…175 °C
Flash Pt: 359.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00151943
CAS Number: 133174-15-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty