Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid 97%

Supplier: Ambeed
Warning
Synonyms: Fmoc-N-Me-Ala-OHN-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-alanineN-Fmoc-N-methyl-L-alanine

A158676-5G A158676-10G A158676-100G A158676-5G A158676-25G A158676-1G A158676-1G
103843-368EA 11 USD InStock
103843-368 103805-154 77286-364 77300-344 77319-228 77286-366 77300-346
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)propanoic acid
Caution: for Research Use Only


Formula: C₁₉H₁₉NO₄
MW: 325.4 g/mol
Boiling Pt: 514.9 °C (760 mm Hg)
Melting Pt: 140 °C
Flash Pt: 265.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00153384
CAS Number: 84000-07-7
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty