Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Alanine benzyl ester hydrochloride 98%

Supplier: Ambeed
Warning
Synonyms: H-Ala-OBzl.HCl

A169435-25G A169435-5G A169435-5G
103844-642EA 30 USD InStock
103844-642 77081-724 77081-722
L-Alanine benzyl ester hydrochloride 98%
L-Alanine benzyl ester hydrochloride
Caution: for Research Use Only


Formula: C₁₀H₁₃NO₂
MW: 215.68 g/mol
Boiling Pt: 256.5 °C (760 mm Hg)
Melting Pt: 140…142 °C
Flash Pt: 119.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00054340
CAS Number: 5557-83-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty