Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-Epigallocatechin gallate 99%

Supplier: Ambeed
Warning
Synonyms: (-)-cis-3,4-Dihydro-5,7-dihydroxy-2-(3,4,5-trihydroxyphenyl)-1(2H)-benzopyran-3-yl Gallate(2R,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoateEGCG

A110153-250MG A110153-1G A110153-5G A110153-100G A110153-25G A110153-10G A110153-100MG A110153-1G
77135-974EA 38.11 USD InStock
77135-974 77135-976 77570-572 77135-972 103797-626 77283-952 77135-980 77135-978
(-)-Epigallocatechin gallate 99%
(-)-Epigallocatechin gallate
Caution: for Research Use Only


Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
Boiling Pt: 909.1 °C (760 mm Hg)
Melting Pt: 216…218 °C
Flash Pt: 320 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075940
CAS Number: 989-51-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty