Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-2-amino-4-chlorobenzoate 98%

Supplier: Ambeed
Warning
Synonyms: Methyl 2-amino-4-chlorobenzoate

A277404-100G A277404-25G A277404-5G A277404-5G
103853-288EA 6 USD InStock
103853-288 77093-188 77093-186 76816-322
Methyl-2-amino-4-chlorobenzoate 98%
Methyl 2-amino-4-chlorobenzoate
Caution: for Research Use Only


Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Boiling Pt: 284 °C (760 mm Hg)
Melting Pt: 66…71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00017568
CAS Number: 5900-58-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty