Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-5-bromovalerate 97%

Supplier: Ambeed
Warning
Synonyms: 5-Bromovaleric acid methyl esterMethyl 5-Bromovalerate

A112439-500G A112439-25G A112439-5G A112439-5G A112439-100G A112439-10G
76718-152EA 408.91 USD InStock
76718-152 77310-654 76718-146 103823-848 76718-148 76718-150
Methyl-5-bromovalerate 97%
Methyl 5-bromovalerate
Caution: for Research Use Only


Formula: BrH₂C(CH₂)₃COOCH₃
MW: 195.06 g/mol
Boiling Pt: 188.2 °C (760 mm Hg)
Flash Pt: 99.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000265
CAS Number: 5454-83-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty