Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nα-Boc-Nε-trifluoroacetyl-L-lysine 98%

Supplier: Ambeed
Warning
Synonyms: N2-Boc-N6-(2,2,2-trifluoroacetyl)-L-lysine

A325185-5G A325185-1G A325185-1G
103836-336EA 17.31 USD InStock
103836-336 103825-394 76839-266
Nα-Boc-Nε-trifluoroacetyl-L-lysine 98%
N-α-Boc-N-φ-trifluoroacetyl-L-lysine
Caution: for Research Use Only


Formula: C₁₃H₂₁F₃N₂O₅
MW: 342.315 g/mol
Boiling Pt: 485.6 °C (760 mm Hg)
Melting Pt: 102…105 °C
Storage Temperature: Freezer
MDL Number: MFCD00037104
CAS Number: 16965-06-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty