Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-glutamic acid 98%

Supplier: Ambeed
Warning

A186619-1000G A186619-500G A186619-5G A186619-100G A186619-25G
77274-326EA 248 USD InStock
77274-326 77084-200 103846-528 77274-328 77274-330
N-Acetyl-L-glutamic acid 98%
N-Acetyl-L-glutamic acid
Caution: for Research Use Only


Formula: C₇H₁₁NO₅
Boiling Pt: 495.9 °C (760 mm Hg)
Melting Pt: 194…196 °C
Flash Pt: 253.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00002802
CAS Number: 1188-37-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty