Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Purine 98%

Supplier: Ambeed
Warning
Synonyms: 1H-Purine

A727505-250MG A727505-10G A727505-5G A727505-5G A727505-100MG A727505-1G A727505-25G
76845-000EA 150.38 USD InStock
76845-000 76727-934 76727-936 76727-938 103840-772 76844-998 76844-996
Purine 98%
Purine
Caution: for Research Use Only


Formula: C₅H₄N₄
MW: 120.11 g/mol
Melting Pt: 214…217 °C
Flash Pt: 225.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00079221
CAS Number: 120-73-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty