Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyrrole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A100308-1G A100308-5G A100308-10G A100308-100G
103821-062EA 12 USD InStock
103821-062 77008-260 103795-356
Pyrrole-2-carboxylic acid 98%
Pyrrole-2-carboxylic acid
Caution: for Research Use Only


Formula: C₅H₅NO₂
MW: 111.10 g/mol
Boiling Pt: 340.3 °C (760 mm Hg)
Melting Pt: 204…208 °C
Flash Pt: 159.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00005219
CAS Number: 634-97-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty